1-(4-(Benzyloxy)-3,5-dimethylphenyl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

303846-1G 303846-500MG 303846-5G
77468-608EA 415.09 USD
77468-608 77481-292 77491-674
1-(4-(Benzyloxy)-3,5-dimethylphenyl)-2-chloro-2,2-difluoroethanone ≥95%
1-(4-(Benzyloxy)-3,5-dimethylphenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₇H₁₅ClF₂O₂
MW: 324.7496 g/mol
Storage Temperature: Ambient
CAS Number: 3002496-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR