1-(4-(Benzyloxy)-3,5-dimethylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

303844-1G 303844-500MG 303844-5G
77468-604EA 415.09 USD
77468-604 77481-288 77491-670
1-(4-(Benzyloxy)-3,5-dimethylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(4-(Benzyloxy)-3,5-dimethylphenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₇H₁₅F₃O₂
MW: 308.2950 g/mol
Storage Temperature: Ambient
CAS Number: 3002473-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR