(4-Isopropoxy-2-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

303834-1G 303834-500MG 303834-5G
77468-600EA 415.09 USD
77468-600 77481-284 77491-666
(4-Isopropoxy-2-methylphenyl)(methyl)sulfane ≥95%
(4-Isopropoxy-2-methylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₆OS
MW: 196.3091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892148
CAS Number: 2969475-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR