1-(4-Isopropoxy-2-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

303832-1G 303832-500MG 303832-5G
77468-596EA 415.09 USD
77468-596 77481-280 77491-662
1-(4-Isopropoxy-2-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(4-Isopropoxy-2-methylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₅H₂₄O₂
MW: 236.3498 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892146
CAS Number: 2969460-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR