Cyclopropyl(4-isopropoxy-2-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

303830-1G 303830-500MG 303830-5G
77468-592EA 415.09 USD
77468-592 77481-276 77491-658
Cyclopropyl(4-isopropoxy-2-methylphenyl)methanol ≥95%
Cyclopropyl(4-isopropoxy-2-methylphenyl)methanol

Precaución: For research use only

Formula: C₁₄H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD34892144
CAS Number: 2969403-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR