2-Chloro-2,2-difluoro-1-(4-isopropoxy-2-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

303828-1G 303828-500MG 303828-5G
77468-588EA 415.09 USD
77468-588 77481-272 77491-654
2-Chloro-2,2-difluoro-1-(4-isopropoxy-2-methylphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(4-isopropoxy-2-methylphenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₃ClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34892142
CAS Number: 2969460-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR