2,2,2-trifluoro-1-(4-isopropoxy-2-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

303826-1G 303826-500MG 303826-5G
77468-584EA 415.09 USD
77468-584 77481-268 77491-650
2,2,2-trifluoro-1-(4-isopropoxy-2-methylphenyl)ethanone ≥95%
2,2,2-trifluoro-1-(4-isopropoxy-2-methylphenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₃F₃O₂
MW: 246.2256 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892140
CAS Number: 2969475-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR