2,2,3,3,3-pentafluoro-1-(2-methoxy-4-methylphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

303786-1G 303786-500MG 303786-5G
77468-580EA 415.09 USD
77468-580 77481-264 77491-646
2,2,3,3,3-pentafluoro-1-(2-methoxy-4-methylphenyl)propan-1-one ≥95%
2,2,3,3,3-pentafluoro-1-(2-methoxy-4-methylphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₁H₉F₅O₂
MW: 268.1799 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892119
CAS Number: 2969459-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR