Trimethyl(3-methyl-5-((3-methylbenzyl)oxy)phenyl)silane ≥95%

Supplier: AOBChem USA

Warning

303783-1G 303783-500MG 303783-5G
77468-576EA 415.09 USD
77468-576 77481-260 77491-642
Trimethyl(3-methyl-5-((3-methylbenzyl)oxy)phenyl)silane ≥95%
Trimethyl(3-methyl-5-((3-methylbenzyl)oxy)phenyl)silane

Precaución: For research use only

Formula: C₁₈H₂₄OSi
Storage Temperature: Ambient
MDL Number: MFCD34892116
CAS Number: 2969205-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR