(3-methyl-5-((3-methylbenzyl)oxy)phenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

303781-1G 303781-500MG 303781-5G
77468-572EA 415.09 USD
77468-572 77481-256 77491-638
(3-methyl-5-((3-methylbenzyl)oxy)phenyl)(phenyl)methanol ≥95%
(3-methyl-5-((3-methylbenzyl)oxy)phenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₂₂H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34892114
CAS Number: 2969459-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR