1-(3-Methyl-5-((3-methylbenzyl)oxy)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

303779-1G 303779-500MG 303779-5G
77468-568EA 415.09 USD
77468-568 77481-252 77491-634
1-(3-Methyl-5-((3-methylbenzyl)oxy)phenyl)cyclobutanol ≥95%
1-(3-Methyl-5-((3-methylbenzyl)oxy)phenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₉H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34892112
CAS Number: 2969403-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR