Methyl(3-Methyl-5-((3-Methylbenzyl)oxy)phenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

303777-1G 303777-500MG 303777-5G
77468-564EA 415.09 USD
77468-564 77481-248 77491-630
Methyl(3-Methyl-5-((3-Methylbenzyl)oxy)phenyl)sulfane ≥95%
Methyl(3-Methyl-5-((3-Methylbenzyl)oxy)phenyl)sulfane

Precaución: For research use only

Formula: C₁₆H₁₈OS
Storage Temperature: Ambient
MDL Number: MFCD34892110
CAS Number: 2969205-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR