1-(3-Methyl-5-((3-methylbenzyl)oxy)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

303774-1G 303774-500MG 303774-5G
77468-560EA 415.09 USD
77468-560 77481-244 77491-626
1-(3-Methyl-5-((3-methylbenzyl)oxy)phenyl)ethanone ≥95%
1-(3-Methyl-5-((3-methylbenzyl)oxy)phenyl)ethanone

Precaución: For research use only

Formula: C₁₇H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD34892107
CAS Number: 2969403-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR