Cyclopropyl(3-methyl-5-((3-methylbenzyl)oxy)phenyl)methanol ≥95%

Supplier: AOBChem USA

303771-1G 303771-500MG 303771-5G
77468-556EA 415.09 USD
77468-556 77481-240 77491-622
Cyclopropyl(3-methyl-5-((3-methylbenzyl)oxy)phenyl)methanol ≥95%
Cyclopropyl(3-methyl-5-((3-methylbenzyl)oxy)phenyl)methanol

Precaución: For research use only

Formula: C₁₉H₂₂O₂
MDL Number: MFCD34892104
CAS Number: 2969475-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR