2,2-Difluoro-1-(3-methyl-5-((3-methylbenzyl)oxy)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

303768-1G 303768-500MG 303768-5G
77468-552EA 415.09 USD
77468-552 77481-236 77491-618
2,2-Difluoro-1-(3-methyl-5-((3-methylbenzyl)oxy)phenyl)ethanone ≥95%
2,2-Difluoro-1-(3-methyl-5-((3-methylbenzyl)oxy)phenyl)ethanone

Precaución: For research use only

Formula: C₁₇H₁₆F₂O₂
MW: 290.3045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892101
CAS Number: 2969403-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR