N,N-Diethyl-4-(methylthio)aniline ≥95%

Supplier: AOBChem USA

Warning

303759-1G 303759-500MG 303759-5G
77468-548EA 347.11 USD
77468-548 77481-232 77491-614
N,N-Diethyl-4-(methylthio)aniline ≥95%
N,N-Diethyl-4-(methylthio)aniline

Precaución: For research use only

Formula: C₁₁H₁₇NS
MW: 195.3243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892094
CAS Number: 21608-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR