N,N-Diethyl-4-(triethylsilyl)aniline ≥95%

Supplier: AOBChem USA

Warning

303754-1G 303754-500MG 303754-5G
77468-544EA 415.09 USD
77468-544 77481-228 77491-610
N,N-Diethyl-4-(triethylsilyl)aniline ≥95%
N,N-Diethyl-4-(triethylsilyl)aniline

Precaución: For research use only

Formula: C₁₆H₂₉NSi
MW: 263.4936 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892091
CAS Number: 2969467-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR