Tributyl(2-methoxyphenyl)stannane ≥95%

Supplier: AOBChem USA

Warning

303180-1G 303180-500MG 303180-5G
77468-532EA 289.43 USD
77468-532 77481-216 77491-598
Tributyl(2-methoxyphenyl)stannane ≥95%
Tributyl(2-methoxyphenyl)stannane

Precaución: For research use only

Formula: C₁₉H₃₄OSn
MW: 397.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01319039
CAS Number: 86487-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR