Ethyl(3-isobutoxy-5-methylphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

302390-1G 302390-500MG 302390-5G
77468-524EA 415.09 USD
77468-524 77481-208 77491-588
Ethyl(3-isobutoxy-5-methylphenyl)sulfane ≥95%
Ethyl(3-isobutoxy-5-methylphenyl)sulfane

Precaución: For research use only

Formula: C₁₃H₂₀OS
Storage Temperature: Ambient
MDL Number: MFCD34891737
CAS Number: 2969461-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR