(3-Isobutoxy-5-methylphenyl)(isopropyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

302386-1G 302386-500MG 302386-5G
77468-520EA 415.09 USD
77468-520 77481-204 77491-584
(3-Isobutoxy-5-methylphenyl)(isopropyl)sulfane ≥95%
(3-Isobutoxy-5-methylphenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₁₄H₂₂OS
MW: 238.3888 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34891733
CAS Number: 2969445-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR