2-Chloro-2,2-difluoro-1-(3-isobutoxy-5-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

302384-1G 302384-500MG 302384-5G
77468-516EA 415.09 USD
77468-516 77481-200 77491-580
2-Chloro-2,2-difluoro-1-(3-isobutoxy-5-methylphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(3-isobutoxy-5-methylphenyl)ethanone

Precaución: For research use only

Formula: C₁₃H₁₅ClF₂O₂
MW: 276.7068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34891731
CAS Number: 2969246-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR