2,2,2-trifluoro-1-(3-isobutoxy-5-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

302382-1G 302382-500MG 302382-5G
77468-512EA 415.09 USD
77468-512 77481-196 77491-576
2,2,2-trifluoro-1-(3-isobutoxy-5-methylphenyl)ethanone ≥95%
2,2,2-trifluoro-1-(3-isobutoxy-5-methylphenyl)ethanone

Precaución: For research use only

Formula: C₁₃H₁₅F₃O₂
MW: 260.2522 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34891729
CAS Number: 2969445-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR