3-Bromo-4-methoxy-5-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

30206-1G 30206-500MG 30206-5G
77468-508EA 451.14 USD
77468-508 77481-192 77491-572
3-Bromo-4-methoxy-5-methylpyridine ≥97%
3-Bromo-4-methoxy-5-methylpyridine

Precaución: For research use only

Formula: C₇H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD18257658
CAS Number: 1256807-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR