Ethyl 3-bromo-5-hydroxybenzoate ≥97%

Supplier: AOBChem USA

Warning

30124-1G 30124-500MG 30124-5G
77468-504EA 166.86 USD
77468-504 77481-188 77491-566
Ethyl 3-bromo-5-hydroxybenzoate ≥97%
Ethyl 3-bromo-5-hydroxybenzoate

Precaución: For research use only

Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD10566802
CAS Number: 870673-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR