(4-phenethoxyphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

300659-1G 300659-500MG 300659-5G
77468-500EA 397.58 USD
77468-500 77481-184 77491-562
(4-phenethoxyphenyl)boronic acid ≥95%
(4-phenethoxyphenyl)boronic acid

Precaución: For research use only

Formula: C₁₄H₁₅BO₃
MW: 242.0781 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11540653
CAS Number: 1029439-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR