(5-Bromothiophen-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

30047-1G 30047-500MG 30047-5G
77468-492EA 284.28 USD
77468-492 77481-176 77491-554
(5-Bromothiophen-3-yl)methanol ≥97%
(5-Bromothiophen-3-yl)methanol

Precaución: For research use only

Formula: C₅H₅BrOS
MW: 193.062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12138023
CAS Number: 73919-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR