2-Methyl-1-naphthaldehyde ≥97%

Supplier: AOBChem USA

Warning

29937-1G 29937-500MG 29937-5G
77468-488EA 189.52 USD
77468-488 77481-172 77491-548
2-Methyl-1-naphthaldehyde ≥97%
2-Methyl-1-naphthaldehyde

Precaución: For research use only

Formula: C₁₂H₁₀O
MW: 170.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02261774
CAS Number: 35699-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR