N,N-Dimethyl-2-(methylthio)aniline ≥95%

Supplier: AOBChem USA

Warning

299245-1G 299245-500MG 299245-5G
77468-484EA 415.09 USD
77468-484 77481-168 77491-542
N,N-Dimethyl-2-(methylthio)aniline ≥95%
N,N-Dimethyl-2-(methylthio)aniline

Precaución: For research use only

Formula: C₉H₁₃NS
MW: 167.2712 g/mol
Storage Temperature: Ambient
CAS Number: 2388-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR