2-Chloro-1-(2-(dimethylamino)phenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

299243-1G 299243-500MG 299243-5G
77468-480EA 594.31 USD
77468-480 77481-164 77491-538
2-Chloro-1-(2-(dimethylamino)phenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(2-(dimethylamino)phenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₁₀ClF₂NO
Storage Temperature: Ambient
CAS Number: 1823324-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR