2-(3-(dimethylamino)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

299238-1G 299238-500MG 299238-5G
77468-472EA 668.47 USD
77468-472 77481-156 77491-530
2-(3-(dimethylamino)phenyl)propan-2-ol ≥95%
2-(3-(dimethylamino)phenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₁H₁₇NO
MW: 179.2587 g/mol
Storage Temperature: Ambient
CAS Number: 92546-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR