1-(3-(dimethylamino)phenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

299234-1G 299234-500MG 299234-5G
77468-468EA 434.66 USD
77468-468 77481-152 77491-526
1-(3-(dimethylamino)phenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3-(dimethylamino)phenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₁NO
Storage Temperature: Ambient
CAS Number: 1592581-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR