N,N-Dimethyl-3-(methylthio)aniline ≥95%

Supplier: AOBChem USA

Warning

299232-1G 299232-500MG 299232-5G
77468-464EA 347.11 USD
77468-464 77481-148 77491-522
N,N-Dimethyl-3-(methylthio)aniline ≥95%
N,N-Dimethyl-3-(methylthio)aniline

Precaución: For research use only

Formula: C₉H₁₃NS
MW: 167.2712 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22055909
CAS Number: 2552-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR