3-(2,4-Dimethoxyphenyl)-1-propene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Osmorhizole

298899-1G 298899-500MG 298899-5G
77468-460EA 220.42 USD
77468-460 77481-144 77491-518
3-(2,4-Dimethoxyphenyl)-1-propene ≥95%
3-(2,4-Dimethoxyphenyl)-1-propene

Precaución: For research use only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801175
CAS Number: 3698-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR