4-(1-Naphthalenyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

297902-1G 297902-500MG 297902-5G
77468-456EA 248.23 USD
77468-456 77481-142 77491-514
4-(1-Naphthalenyl)benzaldehyde ≥95%
4-(1-Naphthalenyl)benzaldehyde

Precaución: For research use only

Formula: C₁₇H₁₂O
MW: 232.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04117439
CAS Number: 56432-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR