Isopropyl(3-methyl-5-propoxyphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

297701-1G 297701-500MG 297701-5G
77468-452EA 415.09 USD
77468-452 77481-138 77491-510
Isopropyl(3-methyl-5-propoxyphenyl)sulfane ≥95%
Isopropyl(3-methyl-5-propoxyphenyl)sulfane

Precaución: For research use only

Formula: C₁₃H₂₀OS
Storage Temperature: Ambient
MDL Number: MFCD34890262
CAS Number: 2969464-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR