2-Chloro-2,2-difluoro-1-(3-methyl-5-propoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

297699-1G 297699-500MG 297699-5G
77468-448EA 415.09 USD
77468-448 77481-134 77491-506
2-Chloro-2,2-difluoro-1-(3-methyl-5-propoxyphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(3-methyl-5-propoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₃ClF₂O₂
Storage Temperature: Ambient
CAS Number: 3002469-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR