2-Fluoro-1-(4-isopropoxy-3,5-dimethylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

297696-1G 297696-500MG 297696-5G
77468-444EA 415.09 USD
77468-444 77481-130 77491-502
2-Fluoro-1-(4-isopropoxy-3,5-dimethylphenyl)ethanone ≥95%
2-Fluoro-1-(4-isopropoxy-3,5-dimethylphenyl)ethanone

Precaución: For research use only

Formula: C₁₃H₁₇FO₂
MW: 224.2712 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34890259
CAS Number: 2969361-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR