1-(4-Isopropoxy-3,5-dimethylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

297690-1G 297690-500MG 297690-5G
77468-436EA 415.09 USD
77468-436 77481-122 77491-494
1-(4-Isopropoxy-3,5-dimethylphenyl)cyclobutanol ≥95%
1-(4-Isopropoxy-3,5-dimethylphenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₅H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34890255
CAS Number: 2969419-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR