1-(4-Isopropoxy-3,5-dimethylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

297688-1G 297688-500MG 297688-5G
77468-432EA 415.09 USD
77468-432 77481-118 77491-490
1-(4-Isopropoxy-3,5-dimethylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(4-Isopropoxy-3,5-dimethylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₆H₂₆O₂
MW: 250.3764 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34890254
CAS Number: 2969326-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR