Cyclopropyl(4-isopropoxy-3,5-dimethylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

297686-1G 297686-500MG 297686-5G
77468-428EA 415.09 USD
77468-428 77481-114 77491-486
Cyclopropyl(4-isopropoxy-3,5-dimethylphenyl)methanol ≥95%
Cyclopropyl(4-isopropoxy-3,5-dimethylphenyl)methanol

Precaución: For research use only

Formula: C₁₅H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34890252
CAS Number: 2969361-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR