2,2,3,3,3-pentafluoro-1-(4-isopropoxy-3,5-dimethylphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

297684-1G 297684-500MG 297684-5G
77468-424EA 415.09 USD
77468-424 77481-110 77491-482
2,2,3,3,3-pentafluoro-1-(4-isopropoxy-3,5-dimethylphenyl)propan-1-one ≥95%
2,2,3,3,3-pentafluoro-1-(4-isopropoxy-3,5-dimethylphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₄H₁₅F₅O₂
Storage Temperature: Ambient
CAS Number: 3002531-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR