2,2-Difluoro-1-(4-isopropoxy-3,5-Dimethylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

297682-1G 297682-500MG 297682-5G
77468-420EA 415.09 USD
77468-420 77481-106 77491-478
2,2-Difluoro-1-(4-isopropoxy-3,5-Dimethylphenyl)ethanone ≥95%
2,2-Difluoro-1-(4-isopropoxy-3,5-Dimethylphenyl)ethanone

Precaución: For research use only

Formula: C₁₃H₁₆F₂O₂
MW: 242.2617 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34890250
CAS Number: 2969345-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR