1-(3,5-dimethyl-4-propoxyphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

Warning

297679-1G 297679-500MG 297679-5G
77468-416EA 415.09 USD
77468-416 77481-102 77491-474
1-(3,5-dimethyl-4-propoxyphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(3,5-dimethyl-4-propoxyphenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₄H₁₅F₅O₂
Storage Temperature: Ambient
CAS Number: 3002499-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR