2-Methoxy-4,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

29681-1G 29681-500MG 29681-5G
77468-408EA 229.69 USD
77468-408 77481-094 77491-466
2-Methoxy-4,5-dimethylbenzoic acid ≥97%
2-Methoxy-4,5-dimethylbenzoic acid

Precaución: For research use only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14538071
CAS Number: 91061-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR