4-Bromo-2,3-difluoro-6-iodo-1-(trimethylsilyl)benzene ≥97%

Supplier: AOBChem USA

Warning

29662-1G 29662-500MG 29662-5G
77468-404EA 77.25 USD
77468-404 77481-090 77491-462
4-Bromo-2,3-difluoro-6-iodo-1-(trimethylsilyl)benzene ≥97%
4-Bromo-2,3-difluoro-6-iodo-1-(trimethylsilyl)benzene

Precaución: For research use only

Formula: C₉H₁₀BrF₂ISi
MW: 391.066 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977324
CAS Number: 1807646-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR