(3,5-Dimethyl-4-propoxyphenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

296454-1G 296454-500MG 296454-5G
77468-400EA 415.09 USD
77468-400 77481-086 77491-458
(3,5-Dimethyl-4-propoxyphenyl)triethylsilane ≥95%
(3,5-Dimethyl-4-propoxyphenyl)triethylsilane

Precaución: For research use only

Formula: C₁₇H₃₀OSi
MW: 278.505 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889867
CAS Number: 2969345-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR