(3,5-Dimethyl-4-propoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

296452-1G 296452-500MG 296452-5G
77468-396EA 668.47 USD
77468-396 77481-082 77491-454
(3,5-Dimethyl-4-propoxyphenyl)(methyl)sulfane ≥95%
(3,5-Dimethyl-4-propoxyphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₂H₁₈OS
MW: 210.3357 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22373622
CAS Number: 1443307-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR