1-(3,5-dimethyl-4-propoxyphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

296449-1G 296449-500MG 296449-5G
77468-392EA 415.09 USD
77468-392 77481-078 77491-450
1-(3,5-dimethyl-4-propoxyphenyl)-2,2-difluoroethanone ≥95%
1-(3,5-dimethyl-4-propoxyphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₃H₁₆F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34889863
CAS Number: 2969326-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR