2-(4-(Benzyloxy)-2-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

296445-1G 296445-500MG 296445-5G
77468-388EA 415.09 USD
77468-388 77481-074 77491-446
2-(4-(Benzyloxy)-2-methylphenyl)propan-2-ol ≥95%
2-(4-(Benzyloxy)-2-methylphenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₇H₂₀O₂
MW: 256.3395 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889861
CAS Number: 2969434-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR