(4-(Benzyloxy)-2-methylphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

296441-1G 296441-500MG 296441-5G
77468-384EA 415.09 USD
77468-384 77481-070 77491-442
(4-(Benzyloxy)-2-methylphenyl)(ethyl)sulfane ≥95%
(4-(Benzyloxy)-2-methylphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₁₆H₁₈OS
Storage Temperature: Ambient
CAS Number: 94571-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR